siRNA Details
| Gene Symbol: Shisa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174086.1 | NP_001167557.1 | 3 | 1243 | ||
| NM_028277.2 | NP_082553.2 | 3 | 1243 | |||
| GenBank mRNA | AK049860.1 | - | 3 | 769 | ||
| BC030884.1 | - | 3 | 532 | |||
| GU479981.1 | - | 3 | 772 | |||
| GU479982.1 | - | 3 | 772 | |||

Shisa9
Genomic Map