siRNA Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139140.1 | NP_631879.1 | 2, 3 | 344 | ||
| GenBank mRNA | AB079132.1 | - | 2, 3 | 344 | ||
| AK048720.1 | - | 2, 3 | 386 | |||
| AK135838.1 | - | 2, 3 | 402 | |||
| AK159335.1 | - | 2, 3 | 438 | |||
| AK167788.1 | - | 1, 2 | 21 | |||
| BC072579.1 | - | 2, 3 | 611 | |||

Spats2
Genomic Map