siRNA Details
| Gene Symbol: Zfp444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146024.1 | NP_001139496.1 | 4, 5 | 629 | ||
| NM_028316.3 | NP_082592.2 | 3, 4 | 509 | |||
| GenBank mRNA | AK018079.1 | - | 4, 5 | 630 | ||
| AK089886.1 | - | 3, 4 | 478 | |||
| AK169973.1 | - | 3, 4 | 521 | |||
| BC023396.2 | - | 4, 5 | 695 | |||
| BC037180.1 | - | 3, 4 | 477 | |||
| BC139359.1 | - | 3, 4 | 465 | 8862381 | ||
| BC139361.1 | - | 3, 4 | 465 | 8862383 | ||

Zfp444
Genomic Map