siRNA Details
Gene Symbol: C1qtnf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204152.1 | NP_001191081.1 | 4 | 608 | ||
NM_001204153.1 | NP_001191082.1 | 3 | 563 | |||
NM_028331.5 | NP_082607.3 | 3 | 774 | |||
GenBank mRNA | AK012868.1 | - | 4 | 550 | ||
AK051058.1 | - | 3 | 688 | |||
AK154407.1 | - | 3 | 751 | |||
BC025489.1 | - | 3 | 506 | |||
BC071187.1 | - | 3 | 741 | 6395221 | ||
DQ002399.1 | - | 3 | 740 |
