siRNA Details
| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013290.1 | - | 3 | 246 | ||
| AK015657.1 | - | 4 | 517 | |||
| AK165613.1 | - | 4 | 347 | |||
| BC037766.1 | - | 3 | 221 | |||
| BC054406.1 | - | 3 | 165 | |||
| BI990615.1 | - | 5 | 357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eldr| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013290.1 | - | 3 | 246 | ||
| AK015657.1 | - | 4 | 517 | |||
| AK165613.1 | - | 4 | 347 | |||
| BC037766.1 | - | 3 | 221 | |||
| BC054406.1 | - | 3 | 165 | |||
| BI990615.1 | - | 5 | 357 | |||