siRNA Details
| Gene Symbol: Veph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145820.3 | NP_665819.2 | 10 | 2151 | ||
| GenBank mRNA | AB085187.1 | - | 10 | 2133 | ||
| AB085189.1 | - | 2 | 196 | |||
| AK013398.1 | - | 2 | 211 | |||
| BC075649.1 | - | 10 | 2151 | |||
| BC094339.1 | - | 2 | 164 | |||
| BC128271.1 | - | 9 | 1858 | 40059997 | ||

Veph1
Genomic Map