siRNA Details
| Gene Symbol: 2810459M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144992.1 | NP_001138464.1 | 1, 2 | 714 | ||
| GenBank mRNA | AK013367.1 | - | 1, 2 | 310 | ||
| BC144821.1 | - | 1, 2 | 714 | 9053369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810459M11Rik| Gene Symbol: 2810459M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144992.1 | NP_001138464.1 | 1, 2 | 714 | ||
| GenBank mRNA | AK013367.1 | - | 1, 2 | 310 | ||
| BC144821.1 | - | 1, 2 | 714 | 9053369 | ||