siRNA Details
| Gene Symbol: Ccdc176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028377.3 | NP_082653.2 | 8 | 1389 | ||
| GenBank mRNA | AK133381.1 | - | 8 | 1344 | ||
| BC151099.1 | - | 8 | 1311 | 9088012 | ||
| BC151103.1 | - | 8 | 1311 | 9088016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc176| Gene Symbol: Ccdc176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028377.3 | NP_082653.2 | 8 | 1389 | ||
| GenBank mRNA | AK133381.1 | - | 8 | 1344 | ||
| BC151099.1 | - | 8 | 1311 | 9088012 | ||
| BC151103.1 | - | 8 | 1311 | 9088016 | ||