siRNA Details
| Gene Symbol: 2900008C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013498.1 | - | 1 | 351 | ||
| BC027704.1 | - | 3 | 925 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900008C10Rik| Gene Symbol: 2900008C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013498.1 | - | 1 | 351 | ||
| BC027704.1 | - | 3 | 925 | |||