siRNA Details
| Gene Symbol: Ccnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028399.1 | NP_082675.1 | 9 | 1212 | ||
| GenBank mRNA | AK036772.1 | - | 1160 | |||
| AK047125.1 | - | 5 | 2112 | |||
| AK049863.1 | - | 9 | 1157 | |||
| AK152399.1 | - | 9 | 1160 | |||
| AK154439.1 | - | 9 | 1170 | |||
| AK156231.1 | - | 1181 | ||||
| BC054122.1 | - | 9 | 1212 | 6509024 | ||
| JX965957.1 | - | 9 | 1148 | |||
| JX965958.1 | - | 9 | 1148 | |||

Ccnt2
Genomic Map