siRNA Details
| Gene Symbol: Ccnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028399.1 | NP_082675.1 | 2, 3 | 308 | ||
| GenBank mRNA | AK013634.1 | - | 2, 3 | 255 | ||
| AK036772.1 | - | 256 | ||||
| AK049863.1 | - | 2, 3 | 253 | |||
| AK152399.1 | - | 2, 3 | 256 | |||
| AK154439.1 | - | 2, 3 | 266 | |||
| AK156231.1 | - | 277 | ||||
| AK156621.1 | - | 2, 3 | 264 | |||
| BC054122.1 | - | 2, 3 | 308 | 6509024 | ||
| JX965957.1 | - | 2, 3 | 244 | |||
| JX965958.1 | - | 2, 3 | 244 | |||

Ccnt2
Genomic Map