siRNA Details
| Gene Symbol: Slc17a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182993.2 | NP_892038.2 | 8 | 1135 | ||
| GenBank mRNA | AK159064.1 | - | 8 | 1108 | ||
| BC028938.1 | - | 6 | 574 | |||
| BC054462.1 | - | 6 | 917 | 6494471 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc17a7| Gene Symbol: Slc17a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182993.2 | NP_892038.2 | 8 | 1135 | ||
| GenBank mRNA | AK159064.1 | - | 8 | 1108 | ||
| BC028938.1 | - | 6 | 574 | |||
| BC054462.1 | - | 6 | 917 | 6494471 | ||