siRNA Details
Gene Symbol: Cnih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028408.3 | NP_082684.1 | 3, 4 | 1061 | ||
GenBank mRNA | AK013789.1 | - | 3, 4 | 1058 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cnih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028408.3 | NP_082684.1 | 3, 4 | 1061 | ||
GenBank mRNA | AK013789.1 | - | 3, 4 | 1058 |