siRNA Details
| Gene Symbol: Glrx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028419.2 | NP_082695.1 | 2 | 340 | ||
| GenBank mRNA | AK009158.1 | - | 2 | 345 | ||
| AK013761.1 | - | 2 | 341 | |||
| AK050883.1 | - | 340 | ||||
| AK148847.1 | - | 1 | 626 | |||
| BC050937.1 | - | 2 | 360 | |||
| BC058371.1 | - | 2 | 360 | 6819353 | ||
| DQ083330.1 | - | 2 | 329 | |||

Glrx5
Genomic Map