siRNA Details
| Gene Symbol: Ppil6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028430.1 | NP_082706.1 | 8 | 900 | ||
| GenBank mRNA | AK013818.1 | - | 8 | 862 | ||
| BC147756.1 | - | 8 | 896 | 9056705 | ||
| BC147757.1 | - | 8 | 896 | 9056706 | ||
| BP772786.1 | - | 1 | 217 | |||

Ppil6
Genomic Map