siRNA Details
| Gene Symbol: 2900092C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028434.3 | NP_082710.2 | 4, 5 | 382 | ||
| GenBank mRNA | AK013837.1 | - | 4, 5 | 398 | ||
| BC087883.1 | - | 4, 5 | 382 | 6772737 | ||
| BC099547.1 | - | 4, 5 | 385 | 6741066 | ||

2900092C05Rik
Genomic Map