siRNA Details
| Gene Symbol: Cenpv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028448.1 | NP_082724.1 | 2, 3 | 512 | ||
| GenBank mRNA | AK014046.1 | - | 2, 3 | 512 | ||
| BC147794.1 | - | 2, 3 | 474 | 9087511 | ||
| BC147795.1 | - | 2, 3 | 474 | 9087512 | ||
| BC147835.1 | - | 2, 3 | 474 | 9087552 | ||
| BC147837.1 | - | 2, 3 | 474 | 9087554 | ||

Cenpv
Genomic Map