siRNA Details
| Gene Symbol: Otud3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028453.1 | NP_082729.1 | 4 | 627 | ||
| GenBank mRNA | AK014098.1 | - | 4 | 669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otud3| Gene Symbol: Otud3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028453.1 | NP_082729.1 | 4 | 627 | ||
| GenBank mRNA | AK014098.1 | - | 4 | 669 | ||