siRNA Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028462.1 | NP_082738.1 | 1 | 257 | ||
| GenBank mRNA | AK014242.1 | - | 1 | 135 | ||
| BC049157.1 | - | 1 | 355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fezf1| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028462.1 | NP_082738.1 | 1 | 257 | ||
| GenBank mRNA | AK014242.1 | - | 1 | 135 | ||
| BC049157.1 | - | 1 | 355 | |||