siRNA Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028462.1 | NP_082738.1 | 1 | 731 | ||
| GenBank mRNA | BC049157.1 | - | 1 | 830 | ||
| BC119565.1 | - | 1 | 731 | 40057718 | ||
| BC119566.1 | - | 1 | 731 | 40057721 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fezf1| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028462.1 | NP_082738.1 | 1 | 731 | ||
| GenBank mRNA | BC049157.1 | - | 1 | 830 | ||
| BC119565.1 | - | 1 | 731 | 40057718 | ||
| BC119566.1 | - | 1 | 731 | 40057721 | ||