siRNA Details
| Gene Symbol: Xpot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081056.1 | NP_001074525.1 | 21 | 2920 | ||
| GenBank mRNA | AK014235.1 | - | 4 | 486 | ||
| AK029113.1 | - | 16 | 2372 | |||
| BC094337.1 | - | 6 | 939 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpot| Gene Symbol: Xpot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081056.1 | NP_001074525.1 | 21 | 2920 | ||
| GenBank mRNA | AK014235.1 | - | 4 | 486 | ||
| AK029113.1 | - | 16 | 2372 | |||
| BC094337.1 | - | 6 | 939 | |||