siRNA Details
| Gene Symbol: Prl8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028477.2 | NP_082753.1 | 6 | 755 | ||
| GenBank mRNA | AF525158.1 | - | 6 | 712 | ||
| AK005485.1 | - | 6 | 756 | |||
| BC109185.1 | - | 6 | 754 | 40050251 | ||
| BC109186.1 | - | 6 | 754 | 40050253 | ||

Prl8a1
Genomic Map