siRNA Details
| Gene Symbol: Setd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080793.5 | NP_542983.3 | 4 | 800 | ||
| GenBank mRNA | AF448509.1 | - | 4 | 582 | ||
| AK048924.1 | - | 4 | 637 | |||
| AK129428.1 | - | 4 | 589 | |||
| AK147316.1 | - | 4 | 856 | |||
| AK147413.1 | - | 4 | 626 | |||
| AK147422.1 | - | 4 | 801 | |||
| AK147667.1 | - | 4 | 835 | |||
| AK170161.1 | - | 4 | 644 | |||
| BC050190.1 | - | 4 | 624 | 30007831 | ||

Setd7
Genomic Map