siRNA Details
| Gene Symbol: 1700037C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028484.2 | NP_082760.2 | 3 | 608 | ||
| GenBank mRNA | AK012792.1 | - | 3 | 579 | ||
| AK161085.1 | - | 615 | ||||
| BC147295.1 | - | 3 | 396 | 9056323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700037C18Rik| Gene Symbol: 1700037C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028484.2 | NP_082760.2 | 3 | 608 | ||
| GenBank mRNA | AK012792.1 | - | 3 | 579 | ||
| AK161085.1 | - | 615 | ||||
| BC147295.1 | - | 3 | 396 | 9056323 | ||