siRNA Details
| Gene Symbol: 1700031M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006585.1 | - | 3, 4 | 167 | ||
| AK015131.1 | - | 3, 4 | 168 | |||
| BC048654.1 | - | 3, 4 | 166 | |||
| BC048656.1 | - | 3 | 88 | |||
| BC099572.1 | - | 3, 4 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700031M16Rik| Gene Symbol: 1700031M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006585.1 | - | 3, 4 | 167 | ||
| AK015131.1 | - | 3, 4 | 168 | |||
| BC048654.1 | - | 3, 4 | 166 | |||
| BC048656.1 | - | 3 | 88 | |||
| BC099572.1 | - | 3, 4 | 174 | |||