siRNA Details
| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163427.1 | NP_001156899.1 | 5 | 611 | ||
| NM_028505.2 | NP_082781.1 | 4 | 443 | |||
| GenBank mRNA | AK006658.1 | - | 5 | 577 | ||
| BC115452.2 | - | 4 | 389 | 40100110 | ||

Pradc1
Genomic Map