siRNA Details
| Gene Symbol: Itpripl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163527.1 | NP_001156999.1 | 2 | 1697 | ||
| NM_001163528.1 | NP_001157000.1 | 2 | 1521 | |||
| GenBank mRNA | AK006667.1 | - | 2 | 1515 | ||
| AK028913.1 | - | 3 | 1437 | |||
| BC130218.1 | - | 2 | 1546 | 40141227 | ||

Itpripl1
Genomic Map