siRNA Details
| Gene Symbol: 1700034J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164236.1 | NP_001157708.1 | 2 | 802 | ||
| NM_028509.1 | NP_082785.1 | 2 | 802 | |||
| GenBank mRNA | AK006598.1 | - | 2 | 795 | ||
| AK029686.1 | - | 789 | ||||
| AK076643.1 | - | 2 | 796 | |||
| BC052362.1 | - | 2 | 794 | 6707822 | ||

1700034J05Rik
Genomic Map