siRNA Details
| Gene Symbol: 1700056E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028516.1 | NP_082792.1 | 1 | 807 | ||
| GenBank mRNA | AK006811.1 | - | 1 | 391 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700056E22Rik| Gene Symbol: 1700056E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028516.1 | NP_082792.1 | 1 | 807 | ||
| GenBank mRNA | AK006811.1 | - | 1 | 391 | ||