siRNA Details
| Gene Symbol: Tex33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163612.2 | NP_001157084.1 | 6 | 1008 | ||
| NM_001163613.1 | NP_001157085.1 | 5 | 803 | |||
| NM_028522.1 | NP_082798.1 | 7 | 933 | |||
| GenBank mRNA | AK006856.1 | - | 7 | 907 | ||
| BC100472.1 | - | 7 | 859 | |||
| BC107197.2 | - | 6 | 812 | 40051085 | ||
| BC107198.1 | - | 6 | 812 | 40051089 | ||

Tex33
Genomic Map