siRNA Details
| Gene Symbol: Armt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024261.2 | NP_077223.2 | 5 | 940 | ||
| GenBank mRNA | BC099894.1 | - | 4 | 3696 | ||
| BC138908.1 | - | 5 | 900 | 8861930 | ||
| BC145732.1 | - | 5 | 900 | 40131035 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armt1| Gene Symbol: Armt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024261.2 | NP_077223.2 | 5 | 940 | ||
| GenBank mRNA | BC099894.1 | - | 4 | 3696 | ||
| BC138908.1 | - | 5 | 900 | 8861930 | ||
| BC145732.1 | - | 5 | 900 | 40131035 | ||