siRNA Details
| Gene Symbol: 1700049G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028538.1 | NP_082814.1 | 4 | 2160 | ||
| GenBank mRNA | AK137340.1 | - | 4 | 2160 | ||
| BC141373.1 | - | 4 | 2153 | 9055986 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700049G17Rik| Gene Symbol: 1700049G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028538.1 | NP_082814.1 | 4 | 2160 | ||
| GenBank mRNA | AK137340.1 | - | 4 | 2160 | ||
| BC141373.1 | - | 4 | 2153 | 9055986 | ||