siRNA Details
| Gene Symbol: 1700066B19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033168.2 | NP_001028340.1 | 2 | 134 | ||
| GenBank mRNA | AK006900.1 | - | 360 | |||
| AK162297.1 | - | 2 | 135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700066B19Rik| Gene Symbol: 1700066B19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033168.2 | NP_001028340.1 | 2 | 134 | ||
| GenBank mRNA | AK006900.1 | - | 360 | |||
| AK162297.1 | - | 2 | 135 | |||