siRNA Details
Gene Symbol: Aldh3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028545.2 | NP_082821.2 | 8, 9 | 1226 | ||
GenBank mRNA | AK081788.1 | - | 7, 8 | 1576 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aldh3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028545.2 | NP_082821.2 | 8, 9 | 1226 | ||
GenBank mRNA | AK081788.1 | - | 7, 8 | 1576 |