siRNA Details
| Gene Symbol: Aldh3b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028545.2 | NP_082821.2 | 9 | 1270 | ||
| GenBank mRNA | AK006803.1 | - | 2 | 168 | ||
| AK081788.1 | - | 8 | 1620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh3b3| Gene Symbol: Aldh3b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028545.2 | NP_082821.2 | 9 | 1270 | ||
| GenBank mRNA | AK006803.1 | - | 2 | 168 | ||
| AK081788.1 | - | 8 | 1620 | |||