siRNA Details
| Gene Symbol: Ms4a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198224.3 | NP_937867.2 | 5 | 519 | ||
| GenBank mRNA | AF544402.1 | - | 5 | 524 | ||
| AK006841.1 | - | 5 | 535 | |||
| BC089483.1 | - | 5 | 519 | 6772390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a13| Gene Symbol: Ms4a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198224.3 | NP_937867.2 | 5 | 519 | ||
| GenBank mRNA | AF544402.1 | - | 5 | 524 | ||
| AK006841.1 | - | 5 | 535 | |||
| BC089483.1 | - | 5 | 519 | 6772390 | ||