siRNA Details
Gene Symbol: 1700086O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039796.1 | - | 2 | 461 | ||
AK050707.1 | - | 4 | 826 | |||
AK078369.1 | - | 2 | 557 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700086O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039796.1 | - | 2 | 461 | ||
AK050707.1 | - | 4 | 826 | |||
AK078369.1 | - | 2 | 557 |