siRNA Details
| Gene Symbol: Pebp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028526.4 | NP_082802.1 | 2 | 263 | ||
| NM_028560.4 | NP_082836.2 | 2 | 263 | |||
| GenBank mRNA | AK006750.1 | - | 2 | 264 | ||
| AK006964.1 | - | 2 | 254 | |||
| BC048621.1 | - | 2 | 241 | 6742909 | ||

Pebp4
Genomic Map