siRNA Details
| Gene Symbol: Camkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028576.2 | NP_082852.1 | 8 | 683 | ||
| GenBank mRNA | AK155246.1 | - | 8 | 684 | ||
| BC138272.1 | - | 8 | 831 | 8861291 | ||
| BC138273.1 | - | 8 | 831 | 8861292 | ||
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK147783.1 | - | 721 | |||

Camkmt
Genomic Map