siRNA Details
| Gene Symbol: 1700001O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198000.3 | NP_932117.2 | 5 | 928 | ||
| GenBank mRNA | AK005618.1 | - | 5 | 439 | ||
| AK007200.1 | - | 417 | ||||
| AK076928.1 | - | 5 | 429 | |||
| BC023044.1 | - | 5 | 898 | 5355351 | ||
| CD773886.1 | - | 3 | 556 | |||

1700001O22Rik
Genomic Map