siRNA Details
Gene Symbol: 1700125H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028589.1 | NP_082865.1 | 3 | 249 | ||
GenBank mRNA | BC048674.1 | - | 3 | 249 | 6774125 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700125H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028589.1 | NP_082865.1 | 3 | 249 | ||
GenBank mRNA | BC048674.1 | - | 3 | 249 | 6774125 |