siRNA Details
| Gene Symbol: 2410004P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201332.1 | NP_001188261.1 | 1 | 183 | ||
| NM_001201333.1 | NP_001188262.1 | 1 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410004P03Rik| Gene Symbol: 2410004P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201332.1 | NP_001188261.1 | 1 | 183 | ||
| NM_001201333.1 | NP_001188262.1 | 1 | 183 | |||