siRNA Details
| Gene Symbol: 2410004P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201332.1 | NP_001188261.1 | 2 | 349 | ||
| NM_001201333.1 | NP_001188262.1 | 2 | 349 | |||
| GenBank mRNA | AK010406.1 | - | 2 | 177 | ||
| CX240116.1 | - | 2 | 157 | |||

2410004P03Rik
Genomic Map