siRNA Details
| Gene Symbol: Sult6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163625.1 | NP_001157097.1 | 4 | 504 | ||
| GenBank mRNA | AK010729.1 | - | 2 | 168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult6b1| Gene Symbol: Sult6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163625.1 | NP_001157097.1 | 4 | 504 | ||
| GenBank mRNA | AK010729.1 | - | 2 | 168 | ||