siRNA Details
| Gene Symbol: Ndufaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028611.3 | NP_082887.2 | 10 | 1277 | ||
| GenBank mRNA | AK010752.1 | - | 10 | 1276 | ||
| AK090097.1 | - | 9 | 1683 | |||
| BC028524.1 | - | 3 | 516 | |||
| BC108350.1 | - | 9 | 1671 | 4220568 | ||
| BC116213.1 | - | 3 | 369 | |||
| BC116214.1 | - | 3 | 369 | |||
| BC128473.1 | - | 3 | 369 | |||
| BC128474.1 | - | 3 | 369 | |||

Ndufaf7
Genomic Map