siRNA Details
Gene Symbol: Mvb12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028617.2 | NP_082893.1 | 8, 9 | 791 | ||
GenBank mRNA | AK003644.1 | - | 8, 9 | 771 | ||
BC020119.1 | - | 8, 9 | 791 | 3982577 | ||
BC027419.1 | - | 8, 9 | 767 | 4163554 | ||
BC087734.1 | - | 8, 9 | 788 | 5101734 |
