siRNA Details
Gene Symbol: Whrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008791.2 | NP_001008791.1 | 12, 13 | 3330 | ||
NM_001008792.2 | NP_001008792.1 | 11, 12 | 3309 | |||
NM_001008793.2 | NP_001008793.1 | 11, 12 | 3294 | |||
NM_028640.2 | NP_082916.1 | 11, 12 | 3297 | |||
GenBank mRNA | AK004110.1 | - | 1, 2 | 27 | ||
AK122523.1 | - | 11, 12 | 2425 | |||
AY739114.1 | - | 11, 12 | 3257 | |||
AY739115.1 | - | 12, 13 | 3290 | |||
AY739116.1 | - | 11, 12 | 3269 | |||
AY739117.1 | - | 11, 12 | 3254 | |||
AY739118.1 | - | 8, 9 | 1906 | |||
AY739119.1 | - | 7, 8 | 1873 | |||
AY739120.1 | - | 6, 7 | 1660 | |||
AY739121.1 | - | 6, 7 | 1778 | |||
AY739122.1 | - | 7, 8 | 2110 | |||
BC150757.1 | - | 11, 12 | 2654 | 9087668 |
