siRNA Details
| Gene Symbol: Satl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028655.1 | NP_082931.1 | 6 | 2111 | ||
| GenBank mRNA | AK015086.1 | - | 6 | 1154 | ||
| BC138009.1 | - | 6 | 2111 | 8861028 | ||
| BC138011.1 | - | 6 | 2111 | 8861030 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Satl1| Gene Symbol: Satl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028655.1 | NP_082931.1 | 6 | 2111 | ||
| GenBank mRNA | AK015086.1 | - | 6 | 1154 | ||
| BC138009.1 | - | 6 | 2111 | 8861028 | ||
| BC138011.1 | - | 6 | 2111 | 8861030 | ||