siRNA Details
| Gene Symbol: Fam83e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033170.4 | NP_001028342.2 | 1 | 1385 | ||
| GenBank mRNA | AK144522.1 | - | 1 | 1387 | ||
| BC042793.1 | - | 1 | 232 | |||
| BC139284.1 | - | 1 | 1305 | 8862306 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam83e| Gene Symbol: Fam83e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033170.4 | NP_001028342.2 | 1 | 1385 | ||
| GenBank mRNA | AK144522.1 | - | 1 | 1387 | ||
| BC042793.1 | - | 1 | 232 | |||
| BC139284.1 | - | 1 | 1305 | 8862306 | ||