siRNA Details
| Gene Symbol: Ifitm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053088.2 | NP_444318.1 | 1 | 137 | ||
| GenBank mRNA | AJ009781.1 | - | 1 | 137 | ||
| AK003363.1 | - | 1 | 168 | |||
| BC103790.1 | - | 1 | 137 | 30287396 | ||
| EU380257.1 | - | 1 | 109 | |||
| FJ200451.1 | - | 1 | 189 | |||

Ifitm5
Genomic Map